Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Lambda Human, PE/ATTO 594, Isotype Control, Novus Biologicals™
SDP

Catalog No. NB180025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB180025 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB180025 Supplier Novus Biologicals Supplier No. NBP306875PEATT594
Add to Cart
Add to Cart

Human Polyclonal Isotype Control

IgM Lambda Polyclonal antibody specifically detects IgM Lambda in Human samples. It is validated for Control

Specifications

Antigen IgM Lambda
Classification Polyclonal
Conjugate PE/ATTO 594
Formulation PBS
Host Species Human
Immunogen Human myeloma serum
Quantity 0.1 mL
Regulatory Status RUO
Gene ID (Entrez) 3507
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Type Isotype Controls
Isotype IgM λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.