Learn More
Invitrogen™ Rat IgM Monoclonal Antibody (HIS40), FITC, eBioscience™

Description
Description: The HIS40 monoclonal antibody reacts with the μ heavy chain of rat IgM. It does not react with other classes of rat immunoglobulin including IgD, IgG or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells. Fluorochrome conjugated HIS40 is used as a detection secondary for rat IgM primary antibodies. Applications Reported: The HIS40 antibody has been reported for use in flow cytometric analysis, immunocytochemistry and immunohistochemistry. Applications Tested: The HIS40 antibody has been tested by flow cytometric analysis as a second step to detect rat IgM primary antibodies. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | Rat IgM |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | HIS40 |
| Concentration | 0.5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Igh-6 |
| Gene Accession No. | 0 |
| Gene Alias | Igh-6; IgM; immunoglobulin heavy chain 6; RGD1359202 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.