Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Monoclonal Antibody (II/41), Brilliant Violet™ 605, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$194.00 - $398.00
Specifications
Antigen | IgM |
---|---|
Clone | II/41 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
40-657-9082
![]() |
Invitrogen™
406579082 |
100 μg |
Each for $398.00
|
|
|||||
40-657-9080
![]() |
Invitrogen™
406579080 |
25 μg |
Each for $194.00
|
|
|||||
Description
Mouse IgM is an immunoglobulin M antibody that is produced by B cells in response to an antigen. It is the first antibody produced during an immune response and is the largest immunoglobulin in the serum. Mouse IgM is composed of five heavy chains and five light chains, and it is involved in complement activation and agglutination of antigens. It plays a crucial role in the innate immune response and is important for neutralizing pathogens before the adaptive immune response is fully activated. Mouse IgM is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry.Specifications
IgM | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
AGM1; AI326478; Igh6; Igh-6; Ighm; Igh-M; Igm; immunoglobulin heavy constant mu; MU; muH; VH | |
Ighm | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
II/41 | |
Flow Cytometry | |
Brilliant Violet 605 | |
Rat | |
Mouse | |
P01872 | |
16019 | |
IgG2a κ | |
Affinity chromatography | |
Ighm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title