Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IgM Monoclonal Antibody (II/41), eFluor™ 660, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 50579082
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-579-082 100 μg
50-579-080 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-579-082 Supplier Invitrogen™ Supplier No. 50579082
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

The II/41 MAb reacts with the mu heavy chain of mouse IgM. It does not react with other classes of mouse immunoglobulin including IgD, IgG or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells. Fluorochrome conjugated II/41 can be used as a detection secondary for mouse IgM.

Mouse IgM is an immunoglobulin M antibody that is produced by B cells in response to an antigen. It is the first antibody produced during an immune response and is the largest immunoglobulin in the serum. Mouse IgM is composed of five heavy chains and five light chains, and it is involved in complement activation and agglutination of antigens. It plays a crucial role in the innate immune response and is important for neutralizing pathogens before the adaptive immune response is fully activated. Mouse IgM is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgM
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone II/41
Concentration 0.2 mg/mL
Conjugate eFluor 660
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Ighm
Gene Accession No. P01872
Gene Alias AGM1; AI326478; Igh6; Igh-6; Ighm; Igh-M; Igm; immunoglobulin heavy constant mu; MU; muH; VH
Gene Symbols Ighm
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16019
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
I am unable to analyze my cells stained with eFluor Organic Dyes today. What options do I have?

Our options will depend on the samples you are analyzing.

If cell viability is not critical, you can store your stained samples at 4 degrees C or on ice overnight in the dark and analyze the following day.

For samples stained with eFluor organic fluorochromes, we recommend that cells be suspended in 100 uL of Flow Cytometry Staining Buffer (Cat. No. 00-4222) and 100 uL of eBioscience IC Fixation Buffer (Cat. No. 00-8222); samples can be incubated for up to 3 days at 4 degrees C in the dark. Alternatively, the 1-step Fix/Lyse Solution (Cat. No. 00-5333) can be used. This is a great option when working with whole blood but also works for other cell types.

Can the eFluor Organic fluorochromes be used for intracellular staining?

Yes, the eFluor Organic fluorochromes can be used for intracellular staining. The eFluor organic fluorochromes maintain bright signal and require minimal changes in compensation when fixed with eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) and Permeabilization Buffer (Cat. No. 00-8333-56) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) (as compared to live cells).

Is the eFluor 660 fluorochrome compatible with Anti-Cy5/Alexa Fluor 647 beads?

Yes, in-house studies have demonstrated that the eFluor 660 fluorochrome is recognized by Anti-Cy5/Alexa Fluor 647 beads. Side by side studies with Alexa Fluor 647 versus eFluor 660 conjugated antibodies have demonstrated comparable results.

What buffers are compatible with the eFluor Organic fluorochromes and eVolve QDots?

The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.

What is the difference between eFluor Organic Dyes and eVolve Qdots?

The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.

Are the eFluor Organic Dyes photo-labile?

As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.