Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat IgM Monoclonal Antibody (RM-7B4), PE, eBioscience™
GREENER_CHOICE

Catalog No. 5010776
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-107-76 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-107-76 Supplier Invitrogen™ Supplier No. 12434282
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The monoclonal antibody RM-7B4 recognizes Rat IgGM antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other rat isotypes nor does it crossreact to mouse IgM or any mouse isotype antibodies. Applications Reported: This RM-7B4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RM-7B4 antibody has been tested by flow cytometric analysis of cells stained with a primary rat IgM antibody. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Rat IgM is an immunoglobulin M isotype antibody produced by B cells in rats. It plays a crucial role in the immune system by recognizing and neutralizing antigens, such as bacteria and viruses. Rat IgM has been extensively studied and is widely used in research applications, including immunology, microbiology, and biotechnology. It is commonly used as a control in experiments, as well as a diagnostic tool for detecting infectious diseases and autoimmune disorders. Researchers have developed various techniques for producing and purifying Rat IgM, including hybridoma technology, protein A/G chromatography, and affinity chromatography. Rat IgM is a valuable tool in the field of immunology, and its use has contributed to a better understanding of the immune system and the development of new treatments for various diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat IgM
Applications Flow Cytometry
Classification Monoclonal
Clone RM-7B4
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Igh-6
Gene Accession No. 0
Gene Alias Igh-6; IgM; immunoglobulin heavy chain 6; RGD1359202
Gene Symbols Igh-6
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299357
Target Species Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.