Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Monoclonal Antibody (SA-DA4), Brilliant Ultra Violet™ 395, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
$215.00 - $423.65
Specifications
Antigen | IgM |
---|---|
Clone | SA-DA4 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
36-399-9841
![]() |
Invitrogen™
363999841 |
25 Tests |
Each for $215.00
|
|
|||||
36-399-9842
![]() ![]() |
Invitrogen™
363999842 |
100 Tests |
Each
for $423.65
Save 3% Reg : |
|
|||||
Description
IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.Specifications
IgM | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
AGM1; IGHM; immunoglobulin heavy constant mu; MU; VH | |
IGHM | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
SA-DA4 | |
Flow Cytometry | |
Brilliant Ultraviolet 395 | |
Mouse | |
Human | |
P01871 | |
3507 | |
IgG1 κ | |
Affinity chromatography | |
IGHM |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title