Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IgM Monoclonal Antibody (SA-DA4), Brilliant Violet™ 605, eBioscience™, Invitrogen™

Catalog No. 406999841
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-699-9841 25 Tests
40-699-9842 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-699-9841 Supplier Invitrogen™ Supplier No. 406999841
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgM
Applications Flow Cytometry
Classification Monoclonal
Clone SA-DA4
Concentration 5 μL/Test
Conjugate Brilliant Violet 605
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene IGHM
Gene Accession No. P01871
Gene Alias AGM1; IGHM; immunoglobulin heavy constant mu; MU; VH
Gene Symbols IGHM
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3507
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.