Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Monoclonal Antibody (SA-DA4), eBioscience™
IgM Monoclonal Antibody (SA-DA4), eBioscience™
Supplier: LIFE TECHNOLOGIES 14999882

Description

Specifications
IgM | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P01871 | |
IGHM | |
Affinity chromatography | |
RUO | |
3507 | |
4°C | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen) | |
SA-DA4 | |
Unconjugated | |
IGHM | |
AGM1; IGHM; immunoglobulin heavy constant mu; MU; VH | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction