Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Antibody (2B9cc), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1107091AF405
Description
IgM Monoclonal specifically detects IgM in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
IgM | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IGHM | |
IgM from human serum. | |
0.2 mL | |
B Cell Development and Differentiation Markers, Immunology | |
3507 | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
2B9 | |
Western Blot, Flow Cytometry, ELISA | |
AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Mu-chain of human IgM, Fab - region localized epitope. | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction