Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Mouse anti-Human, BUV805, Clone: UCH-B1 (also known as UCHB1), BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 748927
Description
IgM Monoclonal antibody specifically detects IgM in Human samples. It is validated for Flow Cytometry
Specifications
IgM | |
Monoclonal | |
Brilliant Ultraviolet 805 | |
UCHB1; IGHM; MU | |
Human Prolymphocytic Leukemia B Cells | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
UCH-B1 (also known as UCHB1) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction