Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Mouse, R718, Clone: II/41, Isotype Control, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 752171
Description
The II/41 clone has been reported to react specifically with mouse IgM of Igh-C[a] and Igh-C[b] haplotypes. It has been reported not to react with other Ig isotypes. In addition, the II/41 clone has been reported not to stimulate B-cell proliferation.

Specifications
IgM | |
Monoclonal | |
R718 | |
50 μg | |
Isotype Control (Primary) | |
Liquid |
Flow Cytometry | |
II/41 | |
Mouse | |
RUO | |
Refrigerate Upon Arrival |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction