Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Rat anti-Mouse, Brilliant Violet 711, Clone: R6-60.2, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 747732
Description
IgM Monoclonal antibody specifically detects IgM in Mouse samples. It is validated for Flow Cytometry.
Specifications
IgM | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
IgM | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
R6-60.2 | |
Brilliant Violet 711 | |
50 μg | |
Primary | |
Mouse | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction