Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Rat anti-Mouse, BV750, Clone: II/41, BD Biosciences™
Rat Monoclonal Antibody
Supplier: BD Biosciences 747333
Description
IgM Monoclonal antibody specifically detects IgM in Mouse samples. It is validated for Flow Cytometry
Specifications
IgM | |
Monoclonal | |
Brilliant Violet 750 | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody. |
Flow Cytometry | |
II/41 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Not reported | |
RUO | |
Mouse | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction