Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Rat anti-Mouse, R-PE, Clone: 1B4B1, Southern Biotech™

Rat Monoclonal Antibody
$346.00 - $475.00
Specifications
| Antigen | IgM |
|---|---|
| Clone | 1B4B1 |
| Concentration | 0.1mg/mL |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
Description
SouthernBiotech monoclonal secondary antibodies offer high sensitivity and low background for use in a variety of applications.
Specifications
| IgM | |
| 0.1mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| IgG1 | |
| Store at 2-8°C; Protect from light; Do not freeze |
| 1B4B1 | |
| Flow Cytometry, ELISA | |
| R-PE | |
| RUO | |
| Supplied in PBS/NaN3 and a stabilizing agent | |
| Secondary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title