Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (+)-Igmesine hydrochloride
GREENER_CHOICESDP

Catalog No. 233250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-325-0 50 mg
23-321-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-325-0 Supplier Tocris Bioscience™ Supplier No. 2332/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective sigma1 ligand; antidepressant

(+)-Igmesine hydrochloride is a selective σ1 receptor ligand (KD = 19.1 nM), with little or no activity at σ2 receptors (IC50 > 1000 nM). Inhibits the NMDA-induced increase in cGMP in a concentration-dependent manner (IC50 value is approximately 100 nM). Weak inhibitor of brain 5-HT uptake in vitro. Exhibits neuroprotective and antidepressant effects.

Specifications

Chemical Name or Material (R)-(+)-N-Cyclopropylmethyl-α-ethyl-N-methyl-α-[(2E)-3-phenyl-2-propenyl)benzenemethanamine hydrochloride
CAS 130152-35-1
Assay Percent Range >98%
Quantity 50 mg
Solubility Soluble to 50mM in DMSO
Molecular Formula C23H29N.HCl
Formula Weight Observed MW: 355.94

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.