Learn More
Eisco™ Igneous Rocks Kit
SCI_ED
Catalog No. S24664 Shop All Eisco Products
Change view
Click to view available options
Igneous Rocks Kit
Catalog No.
S24664 Igneous Rocks Kit
1 options
Catalog No. S24664 Supplier Eisco™ Supplier No. ESNG0005
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
For use in classification and identification of Igneous rocks. The Igneous Rocks Kit contains 12 specimens and specimen key.
  • Numbered and stored in sturdy, sectioned container for easy access and safety
  • Average specimen size: 2 to 3 cm
  • Igneous rocks form from cooling and crystallization of magma or lava
  • Can cool slowly underground or be shot hundreds of feet into air
Specifications
Product Type Igneous Rock Kit
For Use With (Application) Study of texture/mineralogy of rock samples
Includes Basalt, diorite, gabbro, granite, pegmatite, obsidian, peridotite, pumice, rhyolite, scoria, syenite, and tuff
Packaging Sectioned container
Quantity 12 Specimens
Specimen Size Approx. 1 in.