Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGSF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542088 Supplier Invitrogen™ Supplier No. PA542088
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LCHGWLQDLV FMLFKEGYAE PVDYQVPTGT MAIFSIDNLT PEDEGVYICR Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 86%; Sheep: 93%.

This gene encodes a member of the immunoglobulin-like domain-containing superfamily. Proteins in this superfamily contain varying numbers of immunoglobulin-like domains and are thought to participate in the regulation of interactions between cells. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGSF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IGSF1
Gene Accession No. Q8N6C5
Gene Alias 5330413N23; 5530402E03; AI747649; betaglycan and inhibin binding protein; CHTE; IGCD1; IGDC1; IgSF1; Immunoglobulin superfamily member 1; immunoglobulin superfamily, member 1; immunoglobulin-like domain-containing protein 1; InhBP; InhBP/p120; inhibin binding protein; inhibin-binding protein; KIAA0364; LOW QUALITY PROTEIN: immunoglobulin superfamily member 1; MGC75490; mKIAA0364; p120; PGSF2; pituitary gland-specific factor 2
Gene Symbols IGSF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human IGSF1 (aa 151-200).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3547
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.