Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGSF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110776
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110776 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110776 Supplier Invitrogen™ Supplier No. PA5110776
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SRISSKFLLL KDKTQMTWIR PSHKTFQVSF LIGALTESNA GLYRCCYWKE TGWSKPSKVL ELEAPGQLPK PIFWIQAETP ALPGCNVNIL CHGWLQDLVF MLFKEGYAEP VDYQVPTGTM AIFSIDNLTP EDEGVYICRT HIQM Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 88%.

This gene encodes a member of the immunoglobulin-like domain-containing superfamily. Proteins in this superfamily contain varying numbers of immunoglobulin-like domains and are thought to participate in the regulation of interactions between cells. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGSF1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IGSF1
Gene Accession No. Q8N6C5
Gene Alias 5330413N23; 5530402E03; AI747649; betaglycan and inhibin binding protein; CHTE; IGCD1; IGDC1; IgSF1; Immunoglobulin superfamily member 1; immunoglobulin superfamily, member 1; immunoglobulin-like domain-containing protein 1; InhBP; InhBP/p120; inhibin binding protein; inhibin-binding protein; KIAA0364; LOW QUALITY PROTEIN: immunoglobulin superfamily member 1; MGC75490; mKIAA0364; p120; PGSF2; pituitary gland-specific factor 2
Gene Symbols IGSF1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human IGSF1. Recombinant protein control fragment (Product #RP-108306).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3547
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.