Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988893
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8893 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8893 Supplier Bioss Supplier No. BS9194R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the immunoglobulin-like domain-containing superfamily. Proteins in this superfamily contain varying numbers of immunoglobulin-like domains and are thought to participate in the regulation of interactions between cells. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen IGSF1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene IGSF1
Gene Accession No. Q8N6C5
Gene Alias 5330413N23; 5530402E03; AI747649; betaglycan and inhibin binding protein; BOS_25226; CHTE; IGCD1; IGDC1; Igsf1; Immunoglobulin superfamily member 1; immunoglobulin superfamily, member 1; Immunoglobulin-like domain-containing protein 1; InhBP; InhBP/p120; inhibin binding protein; Inhibin-binding protein; KIAA0364; MGC75490; mKIAA0364; p120; PGSF2; Pituitary gland-specific factor 2
Gene Symbols IGSF1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IGSF1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3547
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.