Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGSF10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140047
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140047 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140047 Supplier Invitrogen™ Supplier No. PA5140047
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: AQITLPRAEM RPVKHKWTMI SRDNNTKLEH TVLVGGTVGL NCPGQGDPTP HVDWLLADGS KVRAPYVSED GRILIDKSGK LELQMADSFD TGVYHCISSN YDDADILTYR ITVVEPLVEA YQENGIHHTV FIGETLDLPC HSTGIP Highest antigen sequence indentity to the following orthologs: Rat - 75%, Mouse - 79%.

Ig (immunoglobulin) superfamily members exhibit functional characteristics including immune responses, growth factor signaling and cell adhesion. IGSF10 (immunoglobulin superfamily, member 10), also known as Calvaria mechanical force protein 608 (CMF608), is a 2,623 amino acid secreted protein that contains an N-terminal signal peptide, six leucine-rich repeats (LRRs), and 12 immunoglobulin-like repeats. IGSF10 exists as multiple alternatively spliced isoforms, and is expressed in bone. Specifically, expression of IGSF10 is limited to mesenchymal osteochondroprogenitors with fibroblast-like morphology, where it is thought to be involved in the maintenance of the osteochondroprogenitor cells pool and its down-regulation precedes terminal differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGSF10
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IGSF10
Gene Accession No. Q6WRI0
Gene Alias Calvaria mechanical force protein 608; CMF608; IgSF10; Immunoglobulin superfamily member 10; immunoglobulin superfamily, member 10
Gene Symbols IGSF10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IGSF10. Recombinant protein control fragment (Product #RP-90363).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285313
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.