Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGSF11 Monoclonal Antibody (OTI1B9)
GREENER_CHOICE

Catalog No. PIMA526621
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526621 Supplier Invitrogen™ Supplier No. MA526621
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGSF11 Monoclonal Antibody for Western Blot, IHC (P)

Immunoglobulin superfamily member 11 (IGSF11) is a gene located on chromosome 3q13.3, encoding a transmembrane protein that belongs to the immunoglobulin superfamily. IGSF11 is known for its expression in the brain, where it plays a role in neuron-to-neuron adhesion and synaptic organization. The protein is involved in processes such as synapse formation and stabilization, impacting neural connectivity and signaling pathways. IGSF11 is also expressed in other tissues, including the gastrointestinal tract, where it can contribute to cell adhesion functions. Research has linked IGSF11 to cancer biology, as its expression is altered in various cancers, potentially influencing tumor cell adhesion and metastasis. Its role in modulating cell signaling pathways makes it an interesting target for therapeutic interventions in both neurological disorders and cancer. Current studies are focusing on its functional implications in synaptic plasticity and tumor progression, providing valuable insights into IGSF11's diverse biological roles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGSF11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1B9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Igsf11
Gene Accession No. Q5DX21
Gene Alias 1700025L02Rik; brain and testis-specific immunoglobin superfamily protein; brain and testis-specific immunoglobulin superfamily protein; BTIGSF; Bt-IGSF; cancer/testis antigen 119; CT119; CXADR like 1; CXADRL1; Igsf11; Igsf13; Immunoglobulin superfamily member 11; immunoglobulin superfamily, member 11; immunoglobulin superfamily, member 13; V-set and immunoglobulin domain containing 3; V-set and immunoglobulin domain-containing protein 3; VSIG3
Gene Symbols Igsf11
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 23-241 of IGSF11 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152404
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.