Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF11 Mouse anti-Human, Clone: OTI2H1, liquid, TrueMAB™
SDP

Catalog No. 501679925
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-9925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-9925 Supplier OriGene Supplier No. TA803797
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Functions as a cell adhesion molecule through homophilic interaction. Stimulates cell growth.

Specifications

Antigen IGSF11
Applications Western Blot
Classification Monoclonal
Clone OTI2H1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene IGSF11
Gene Accession No. Q5DX21
Gene Alias BT-IgSF, CT119, CXADRL1, Igsf13, VSIG3
Gene Symbols IGSF11
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 23-241 of human IGSF11 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152404
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.