Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGSF11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112964
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112964 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112964 Supplier Invitrogen™ Supplier No. PA5112964
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IGSF11 Polyclonal Antibody for Western Blot, ELISA

Immunoglobulin superfamily member 11 (IGSF11) is a gene located on chromosome 3q13.3, encoding a transmembrane protein that belongs to the immunoglobulin superfamily. IGSF11 is known for its expression in the brain, where it plays a role in neuron-to-neuron adhesion and synaptic organization. The protein is involved in processes such as synapse formation and stabilization, impacting neural connectivity and signaling pathways. IGSF11 is also expressed in other tissues, including the gastrointestinal tract, where it can contribute to cell adhesion functions. Research has linked IGSF11 to cancer biology, as its expression is altered in various cancers, potentially influencing tumor cell adhesion and metastasis. Its role in modulating cell signaling pathways makes it an interesting target for therapeutic interventions in both neurological disorders and cancer. Current studies are focusing on its functional implications in synaptic plasticity and tumor progression, providing valuable insights into IGSF11's diverse biological roles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGSF11
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Igsf11
Gene Accession No. P0C673, Q5DX21
Gene Alias 1700025L02Rik; brain and testis-specific immunoglobin superfamily protein; brain and testis-specific immunoglobulin superfamily protein; BTIGSF; Bt-IGSF; cancer/testis antigen 119; CT119; CXADR like 1; CXADRL1; Igsf11; Igsf13; Immunoglobulin superfamily member 11; immunoglobulin superfamily, member 11; immunoglobulin superfamily, member 13; V-set and immunoglobulin domain containing 3; V-set and immunoglobulin domain-containing protein 3; VSIG3
Gene Symbols Igsf11
Host Species Rabbit
Immunogen Recombinant Human Immunoglobulin superfamily member 11 protein (23-241AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152404, 207683
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.