Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF11 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988898
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8898 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8898 Supplier Bioss Supplier No. BS9201R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Functions as a cell adhesion molecule through homophilic interaction. Stimulates cell growth.

Specifications

Antigen IGSF11
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Igsf11
Gene Accession No. Q5DX21
Gene Alias 1700025L02Rik; brain and testis-specific immunoglobin superfamily protein; Brain and testis-specific immunoglobulin superfamily protein; BTIGSF; Bt-IGSF; cancer/testis antigen 119; CT119; CXADR like 1; CXADRL1; IGSF11; Igsf13; Immunoglobulin superfamily member 11; immunoglobulin superfamily, member 11; immunoglobulin superfamily, member 13; V-set and immunoglobulin domain containing 3; V-set and immunoglobulin domain-containing protein 3; VSIG3
Gene Symbols Igsf11
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IGSF11.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152404
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.