Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGSF2/CD101 Antibody (307707) [DyLight 650], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB3368W
Description
IGSF2/CD101 Monoclonal antibody specifically detects IGSF2/CD101 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
IGSF2/CD101 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9398 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, CyTOF | |
307707 | |
Flow Cytometry, CyTOF-ready | |
CD101 antigen, CD101 molecule, Cell surface glycoprotein V7, EWI101, EWI-101, Glu-Trp-Ile EWI motif-containing protein 101, IgSF2, leukocyte surface protein, member 2 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse IGSF2/CD101, Gln21-Tyr1033, Accession # A8E0Y8 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction