Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF2 Mouse anti-Human, Clone:BB27, Abnova™

Catalog No. 89333053 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-053 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-053 Supplier Abnova Corporation Supplier No. MAB16946
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human IGSF2.

Specifications

Antigen IGSF2
Applications Flow Cytometry
Classification Monoclonal
Clone BB27
Conjugate Unconjugated
Description immunoglobulin superfamily, member 2
Dilution Flow Cytometry
Immunoprecipitation
Immunohistochemistry (Frozen sections)
Western Blot
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (15 mM sodium azide)
Gene IGSF2
Gene Alias CD101, V7
Host Species Mouse
Immunogen Human thymic clone B12.
Quantity 100 μg
Gene ID (Entrez) 9398
Test Specificity This antibody recognizes CD101, a 140 kDa disulfide-bonded homodimeric protein expressed on activated T cells, and some ther cell types, such as granulocytes and cells of the monocyte/macropgage lineage.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.