Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF21 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988901
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8901 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8901 Supplier Bioss Supplier No. BS9204R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein which has two immunoglobulin (Ig) domains and is a member of the immunoglobulin superfamily. Proteins in this superfamily are usually found on or in cell membranes and act as receptors in immune response pathways. The protein is involved in synaptic inhibition in the brain and selectively regulates inhibitory presynaptic differentiation through interacting with presynaptic NRXN2.

Specifications

Antigen IGSF21
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene IGSF21
Gene Accession No. Q96ID5
Gene Alias BC055811; Gm141; Igsf21; immunoglobin superfamily member 21; immunoglobin superfamily, member 21; Immunoglobulin superfamily member 21; immunoglobulin superfamily, member 21; RGD1310117
Gene Symbols IGSF21
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IGSF21.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84966
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.