Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGSF22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571123
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571123 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571123 Supplier Invitrogen™ Supplier No. PA571123
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 100%; Human: 100%.

IGSF22 gene ontology annotations related to this gene include cardiac muscle fiber development; cardiac muscle tissue morphogenesis; cardiac myofibril assembly; muscle contraction; sarcomere organization; skeletal muscle myosin thick filament assembly; skeletal muscle thin filament assembly; striated muscle myosin thick filament assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGSF22
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IGSF22
Gene Accession No. Q8N9C0
Gene Alias IGFN2; IGSF22; Immunoglobulin superfamily member 22; immunoglobulin superfamily, member 22
Gene Symbols IGSF22
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human IGSF22.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283284
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.