Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGSF3 Antibody (002), DyLight 405 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289880V
Description
IGSF3 Monoclonal antibody specifically detects IGSF3 in Human samples. It is validated for ELISASpecifications
IGSF3 | |
Monoclonal | |
DyLight 405 | |
EWI-3, glu-Trp-Ile EWI motif-containing protein 3, immunoglobulin superfamily, member 3, member 3, V8 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IGSF3 (O75054-1; Met1-Ala1124). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
002 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
3321 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction