Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGSF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583319
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583319 Supplier Invitrogen™ Supplier No. PA583319
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGTVEFHDLV TFTRDGGVQW GDRSSSFRTR TAIEKAESSN NVRLSISRAS DTEAGKYQCV AELWRKNYNN TWTRLAERTS NLLEIRVLQP VTKLQVSKSK RTLTLVENKP IQLNCSVKSQ TSQNSHFAVL WYVHKPSDA Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 96%.

IGSF3 (immunoglobulin superfamily, member 3), also known as V8 or EWI-3, is a 1,214 amino acid protein. Widely expressed with predominant expression in kidney, placenta and lung, IGSF3 localizes to the membrane and contains an N-terminal signal peptide, eight immunoglobulin (Ig) domains and a transmembrane segment. IGSF3 exhibits strong sequence and structural similarity to CD101 (32% identity), a leukocyte surface protein with seven Ig domains that is believed to play a role in T-cell activation. Despite the structural similarities between IGSF3 and CD101, IGSF3 is not expressed in peripheral blood lymphocytes and does not appear to participate in an immune function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGSF3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IGSF3
Gene Accession No. O75054
Gene Alias 1700016K10Rik; 2810035F16Rik; 4833439O17Rik; EWI3; EWI-3; Glu-Trp-Ile EWI motif-containing protein 3; Igsf3; immunoglobin superfamily, member 3; immunoglobulin superfamily member 3; immunoglobulin superfamily, member 3; Kiaa0466; LCDD; mKIAA0466; RP4-781D12.1; V8
Gene Symbols IGSF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IGSF3. Recombinant protein control fragment (Product #RP-102708).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3321
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.