Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF4A/SynCAM1/CADM1 Antibody (41N3D3), Novus Biologicals™
SDP

Rat Monoclonal Antibody

$206.50 - $501.50

Specifications

Antigen IGSF4A/SynCAM1/CADM1
Clone 41N3D3
Concentration 0.5 mg/mL
Dilution Western Blot 1-5 ug/ml
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP214841SS
SDP
View Documents
Novus Biologicals
NBP214841SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP214841
SDP
View Documents
Novus Biologicals
NBP214841
0.1 mg
Each for $501.50
Only null left
Add to Cart
 
Description

Description

IGSF4A/SynCAM1/CADM1 Monoclonal specifically detects IGSF4A/SynCAM1/CADM1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

IGSF4A/SynCAM1/CADM1
0.5 mg/mL
Western Blot
Unconjugated
Rat
Growth and Development, Neuronal Cell Markers, Neuroscience
Q9BY67
23705
A portion of amino acids 150-200 of human CADM1 was used as the immunogen for this antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
41N3D3
Western Blot 1-5 ug/ml
Monoclonal
Purified
RUO
Human
BL2, cell adhesion molecule 1, IgSF4, IGSF4ADKFZp686F1789, IGSF4immunoglobulin superfamily, member 4D variant 2, Immunoglobulin superfamily member 4, immunoglobulin superfamily, member 4, Necl-2, NECL2MGC149785, nectin-like 2, Nectin-like protein 2, RA175, sgIGSF, Spermatogenic immunoglobulin superfamily, ST17, sTSLC-1, Synaptic cell adhesion molecule, SynCAM, synCAM1, SYNCAMMGC51880, TSLC1, TSLC-1, TSLC1/Nectin-like 2/IGSF4, Tumor suppressor in lung cancer 1SYNCAM1
CADM1
IgG1 κ
Protein G purified
The amino acid sequence used as immunogen for this antibody is 100% homologous in Human and Primates. 98% in Mouse, Dog, Rabbit, Horse and Chinese Hamster.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.