Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGSF4A/SynCAM1/CADM1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299730APC
Description
IGSF4A/SynCAM1/CADM1 Polyclonal antibody specifically detects IGSF4A/SynCAM1/CADM1 in Human samples. It is validated for ELISASpecifications
IGSF4A/SynCAM1/CADM1 | |
Polyclonal | |
ELISA | |
BL2, cell adhesion molecule 1, IgSF4, IGSF4ADKFZp686F1789, IGSF4immunoglobulin superfamily, member 4D variant 2, Immunoglobulin superfamily member 4, immunoglobulin superfamily, member 4, Necl-2, NECL2MGC149785, nectin-like 2, Nectin-like protein 2, RA175, sgIGSF, Spermatogenic immunoglobulin superfamily, ST17, sTSLC-1, Synaptic cell adhesion molecule, SynCAM, synCAM1, SYNCAMMGC51880, TSLC1, TSLC-1, TSLC1/Nectin-like 2/IGSF4, Tumor suppressor in lung cancer 1SYNCAM1 | |
Produced in rabbits immunized with purified, recombinant Human IGSF4A/SynCAM1/CADM1 (Accession#: NP_055148.3; Met 1-His 374) | |
0.1 mL | |
Growth and Development, Neuronal Cell Markers, Neuroscience | |
23705 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction