Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF4A/SynCAM1/CADM1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$206.50 - $499.50

Specifications

Antigen IGSF4A/SynCAM1/CADM1
Dilution Western Blot 1:1000, Simple Western 1:1000, Immunocytochemistry/Immunofluorescence 1:2000
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB300186SS
SDP
View Documents
Novus Biologicals
NB300186SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB300186
SDP
View Documents
Novus Biologicals
NB300186
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

IGSF4A/SynCAM1/CADM1 Polyclonal specifically detects IGSF4A/SynCAM1/CADM1 in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

IGSF4A/SynCAM1/CADM1
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
Rabbit
Growth and Development, Neuronal Cell Markers, Neuroscience
Whole antisera with 0.02% Sodium Azide
BL2, cell adhesion molecule 1, IgSF4, IGSF4ADKFZp686F1789, IGSF4immunoglobulin superfamily, member 4D variant 2, Immunoglobulin superfamily member 4, immunoglobulin superfamily, member 4, Necl-2, NECL2MGC149785, nectin-like 2, Nectin-like protein 2, RA175, sgIGSF, Spermatogenic immunoglobulin superfamily, ST17, sTSLC-1, Synaptic cell adhesion molecule, SynCAM, synCAM1, SYNCAMMGC51880, TSLC1, TSLC-1, TSLC1/Nectin-like 2/IGSF4, Tumor suppressor in lung cancer 1SYNCAM1
CADM1
IgG
Unpurified
Western Blot 1:1000, Simple Western 1:1000, Immunocytochemistry/Immunofluorescence 1:2000
Polyclonal
Antisera
RUO
Human, Rat
Q9BY67
23705
A synthetic peptide corresponding to the C-terminal region of rat SynCAM/CADM1.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.