Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGSF4B/SynCAM3/CADM3 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298284DL594
Description
IGSF4B/SynCAM3/CADM3 Polyclonal specifically detects IGSF4B/SynCAM3/CADM3 in Mouse samples. It is validated for Western Blot, ELISA.Specifications
IGSF4B/SynCAM3/CADM3 | |
Polyclonal | |
Western Blot, ELISA | |
BIgR, Brain immunoglobulin receptor, cell adhesion molecule 3, IgSF4B, IGSF4BSynCAM3, Immunoglobulin superfamily member 4B, member 4B, NECL-1, NECL1TSLC1-like protein 1, Nectin-like protein 1, Synaptic cell adhesion molecule 3, synCAM3, TSLC1-like 1, TSLL1SYNCAM3 | |
Produced in rabbits immunized with purified, recombinant IGSF4B/SynCAM3/CADM3 (NP_444429.1; Met1-His328). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot, ELISA | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
57863 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction