Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGSF4C/SynCAM4 Antibody (404619), DyLight 550, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB4164L
Description
IGSF4C/SynCAM4 Monoclonal antibody specifically detects IGSF4C/SynCAM4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
IGSF4C/SynCAM4 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
199731 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
404619 | |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
cell adhesion molecule 4, IgSF4C, IGSF4Cnectin-like 4, Immunoglobulin superfamily member 4C, immunoglobulin superfamily, member 4C, Necl-4, NECL4synCAM4, Nectin-like protein 4, SYNCAM4, TSLC1-like 2, TSLC1-like protein 2, TSLL2SynCAM4 | |
Mouse myeloma cell line NS0-derived recombinant human IGSF4C/SynCAM4, Gln25-Tyr323, Accession # Q8NFZ8 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction