Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGSF4C/SynCAM4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP314360
This item is not returnable.
View return policy
Description
IGSF4C/SynCAM4 Polyclonal antibody specifically detects IGSF4C/SynCAM4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
IGSF4C/SynCAM4 | |
Polyclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
199731 | |
Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
cell adhesion molecule 4, IgSF4C, IGSF4Cnectin-like 4, Immunoglobulin superfamily member 4C, immunoglobulin superfamily, member 4C, Necl-4, NECL4synCAM4, Nectin-like protein 4, SYNCAM4, TSLC1-like 2, TSLC1-like protein 2, TSLL2SynCAM4 | |
Synthetic 14 amino acid peptide from internal region of human IGSF4C/SynCAM4. | |
50 μg | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction