Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGSF5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547542
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547542 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547542 Supplier Invitrogen™ Supplier No. PA547542
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse (rm) JAM-A, rmJAM-B, and rmJAM-C is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Provides, together with MAGI1, an adhesion machinery at tight junctions, which may regulate the permeability of kidney glomerulus and small intestinal epithelial cells. Mediates calcium-independent homophilic cell adhesion. In testis, it may function as a cell adhesion molecule rather than a tight-junction protein. It may participate in the adhesion between spermatogonia-spermatogonia, spermatogonia-Sertoli cells, and Sertoli cells-Sertoli cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGSF5
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene IGSF5
Gene Accession No. Q7TSN7
Gene Alias 2010003D20Rik; cell adhesion molecule JCAM; GSF5; IgSF5; immunoglobulin superfamily 5 like; immunoglobulin superfamily member 5; immunoglobulin superfamily, member 5; Jam4; JAM-4; JCAM; junction adhesion molecule 4; junctional adhesion molecule 4
Gene Symbols IGSF5
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse JAM-4/IGSF5 Leu15-Ala139.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72058
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.