Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IGSF5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580713
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human IGSF5.
Provides, together with MAGI1, an adhesion machinery at tight junctions, which may regulate the permeability of kidney glomerulus and small intestinal epithelial cells. Mediates calcium-independent homophilic cell adhesion. In testis, it may function as a cell adhesion molecule rather than a tight-junction protein. It may participate in the adhesion between spermatogonia-spermatogonia, spermatogonia-Sertoli cells, and Sertoli cells-Sertoli cells.
Specifications
IGSF5 | |
Polyclonal | |
Unconjugated | |
IGSF5 | |
2010003D20Rik; cell adhesion molecule JCAM; GSF5; IgSF5; immunoglobulin superfamily 5 like; immunoglobulin superfamily member 5; immunoglobulin superfamily, member 5; Jam4; JAM-4; JCAM; junction adhesion molecule 4; junctional adhesion molecule 4 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
150084 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Q9NSI5 | |
IGSF5 | |
A synthetic peptide corresponding to the C-terminus of the Human IGSF5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction