Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGSF5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581411
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581411 Supplier Invitrogen™ Supplier No. PA581411
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has specificity for Mouse IGSF5.

Provides, together with MAGI1, an adhesion machinery at tight junctions, which may regulate the permeability of kidney glomerulus and small intestinal epithelial cells. Mediates calcium-independent homophilic cell adhesion. In testis, it may function as a cell adhesion molecule rather than a tight-junction protein. It may participate in the adhesion between spermatogonia-spermatogonia, spermatogonia-Sertoli cells, and Sertoli cells-Sertoli cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGSF5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7
Gene IGSF5
Gene Accession No. Q7TSN7
Gene Alias 2010003D20Rik; cell adhesion molecule JCAM; GSF5; IgSF5; immunoglobulin superfamily 5 like; immunoglobulin superfamily member 5; immunoglobulin superfamily, member 5; Jam4; JAM-4; JCAM; junction adhesion molecule 4; junctional adhesion molecule 4
Gene Symbols IGSF5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Mouse IGSF5.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72058
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.