Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988895
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8895 Supplier Bioss Supplier No. BS9197R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Provides, together with MAGI1, an adhesion machinery at tight junctions, which may regulate the permeability of kidney glomerulus and small intestinal epithelial cells. Mediates calcium-independent homophilic cell adhesion. In testis, it may function as a cell adhesion molecule rather than a tight-junction protein. It may participate in the adhesion between spermatogonia-spermatogonia, spermatogonia-Sertoli cells, and Sertoli cells-Sertoli cells.

Specifications

Antigen IGSF5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene IGSF5
Gene Accession No. Q9NSI5
Gene Alias 2010003D20Rik; cell adhesion molecule JCAM; GSF5; IGSF5; immunoglobulin superfamily 5 like; Immunoglobulin superfamily member 5; immunoglobulin superfamily, member 5; JAM4; JAM-4; JCAM; junction adhesion molecule 4; Junctional adhesion molecule 4
Gene Symbols IGSF5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IGSF5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150084
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.