Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGSF6/DORA Antibody (104A10.01), Alexa Fluor™ 546, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals DDX0220A546100
Description
IGSF6/DORA Monoclonal specifically detects IGSF6/DORA in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry.Specifications
IGSF6/DORA | |
Monoclonal | |
0.5mg/mL | |
Western Blot, Flow Cytometry 1:10-1:1000 | |
IGSF6 | |
DORA/IgSF6 was used as immunogen. | |
0.1 mg | |
Primary | |
Human, Mouse, Canine | |
IgG1 |
Western Blot, Flow Cytometry | |
104A10.01 | |
Alexa Fluor 546 | |
DORAdown-regulated by activation (immunoglobulin superfamily), IgSF6, immunoglobulin superfamily member 6, immunoglobulin superfamily, member 6, Protein DORA | |
Mouse | |
Ion exchange chromatography | |
RUO | |
10261 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction