Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGSF8/CD316 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP314381
This item is not returnable.
View return policy
Description
IGSF8/CD316 Polyclonal antibody specifically detects IGSF8/CD316 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
IGSF8/CD316 | |
Polyclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Antibody |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.6 mg/mL | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CD316, CD316 antigen, CD81P3LIR-D1, EWI-2, EWI2CD81 partner 3, Glu-Trp-Ile EWI motif-containing protein 2, IgSF8, immunoglobulin superfamily, member 8, KCT4, KCT-4, Keratinocytes-associated transmembrane protein 4, PGRLimmunoglobulin superfamily member 8 | |
Synthetic 15 amino acid peptide from internal region of human IGSF8. | |
50 μg | |
Neuroscience | |
93185 | |
Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction