Learn More
Invitrogen™ IGSF8 Polyclonal Antibody

Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Monkey, Galago, Rabbit (100%) Panda, Bovine, Bat, Pig, Guinea pig (93%).
Specifications
Specifications
| Antigen | IGSF8 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | IGSF8 |
| Gene Accession No. | Q969P0 |
| Gene Alias | AA033172; AU041109; CD316; CD81 partner 3; CD81P3; ESTM34; EWI2; EWI-2; Glu-Trp-Ile EWI motif-containing protein 2; IGSF8; Immunoglobulin superfamily member 8; immunoglobulin superfamily, member 8; Kct4; KCT-4; Keratinocyte-associated transmembrane protein 4; keratinocytes-associated transmembrane protein 4; LIR-D1; PG regulatory-like protein; PGRL; Prostaglandin regulatory-like protein; R75384 |
| Gene Symbols | IGSF8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.