Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGSF8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532874
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532874 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532874 Supplier Invitrogen™ Supplier No. PA532874
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Bovine, Bat, Elephant, Panda, Rabbit, Pig (100%); Mouse, Rat (93%). Immunohistochemistry with formalin-fixed paraffin-embedded tissues requires pretreatment using HIER with citrate buffer.

This gene encodes a member the EWI subfamily of the immunoglobulin protein superfamily. Members of this family contain a single transmembrane domain, an EWI (Glu-Trp-Ile)-motif and a variable number of immunoglobulin domains. This protein interacts with the tetraspanins CD81 and CD9 and may regulate their role in certain cellular functions including cell migration and viral infection. The encoded protein may also function as a tumor suppressor by inhibiting the proliferation of certain cancers. Alternate splicing results in multiple transcript variants that encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGSF8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene IGSF8
Gene Accession No. Q969P0
Gene Alias AA033172; AU041109; CD316; CD81 partner 3; CD81P3; ESTM34; EWI2; EWI-2; Glu-Trp-Ile EWI motif-containing protein 2; IGSF8; Immunoglobulin superfamily member 8; immunoglobulin superfamily, member 8; Kct4; KCT-4; Keratinocyte-associated transmembrane protein 4; keratinocytes-associated transmembrane protein 4; LIR-D1; PG regulatory-like protein; PGRL; Prostaglandin regulatory-like protein; R75384
Gene Symbols IGSF8
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from internal region of human IGSF8.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100154024, 100338663, 519266, 93185
Target Species Human, Rabbit, Monkey, Bovine, Pig, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.