Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IGSF9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558089
Description
Immunogen sequence: TPLPIGMPGV IRCPVRANPP LLFVSWTKDG KALQLDKFPG WSQGTEGSLI IALGNEDALG EYSCTPYNSL GTAGPSPVTR VLLKAPPAFI ERPKEEYFQE VG Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.

Specifications
IGSF9 | |
Polyclonal | |
Unconjugated | |
IGSF9 | |
644ETD8; Dasm1; Dendrite arborization and synapse maturation protein 1; FP18798; Igsf9; Igsf9a; immunoglobulin superfamily member 9; immunoglobulin superfamily member 9A; immunoglobulin superfamily, member 9; KIAA1355; Kiaa1355-hp; mKIAA1355; Ncaml; NCAM-like protein NRT1; Nrt1; protein turtle homolog A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57549 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2J2 | |
IGSF9 | |
Recombinant protein corresponding to Human IGSF9. Recombinant protein control fragment (Product #RP-96545). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction