Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGSF9B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552742
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552742 Supplier Invitrogen™ Supplier No. PA552742
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEAPLSSGKV SPESIRTLRA PSESSDDQGQ PAAKRMLSPT REKELSLYKK TKRAISSKKY SVAKAEAEAE ATTPIELISR GPDGRFVMDP AEMEPSLKSR RIEGFPFAEE TDMYPEFRQS DEENEDPLVP TSVAALKSQL TPLSSSQE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

IGSF9B is a protein coding gene. Transmembrane protein which is abundantly expressed in interneurons, where it may regulate inhibitory synapse development. May mediate homophilic cell adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGSF9B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IGSF9B
Gene Accession No. Q9UPX0
Gene Alias Gm508; IgSF9B; igsf9l; igsf9l protein; im:7148508; immunoglobulin superfamily member 9B; immunoglobulin superfamily, member 9b; immunoglobulin superfamily, member 9b; protein turtle homolog A; Kiaa1030; LOW QUALITY PROTEIN: protein turtle homolog B; mKIAA1030; Protein turtle homolog B; RGD1564717; si:dkey-49h9.6
Gene Symbols IGSF9B
Host Species Rabbit
Immunogen Recombinant protein fragment of Human IGSF9B (Product #RP-89998)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22997
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.