Learn More
Invitrogen™ IGSF9B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564934
Description
Immunogen sequence: LEAPLSSGKVS PESIRTLRAP SESSDDQGQP AAKRMLSPTR EKELSLYKKT KRAISSKKYS VAKAEAEAEA TTPIELISRG PDGRFVMDPA EMEPSLKSRR IEGFPFAEET DMYPEFRQSD EENEDPLVPT SVAALKSQLT PLSSSQE Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
IGSF9B is a protein coding gene. Transmembrane protein which is abundantly expressed in interneurons, where it may regulate inhibitory synapse development. May mediate homophilic cell adhesion.
Specifications
IGSF9B | |
Polyclonal | |
Unconjugated | |
IGSF9B | |
Gm508; IgSF9B; igsf9l; igsf9l protein; im:7148508; immunoglobulin superfamily member 9B; immunoglobulin superfamily, member 9b; immunoglobulin superfamily, member 9b; protein turtle homolog A; Kiaa1030; LOW QUALITY PROTEIN: protein turtle homolog B; mKIAA1030; Protein turtle homolog B; RGD1564717; si:dkey-49h9.6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22997 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UPX0 | |
IGSF9B | |
Recombinant Human IGSF9B. Recombinant protein control fragment (Product #RP-89998). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.