Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgT Antibody (Z55F8*C3), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261127AF350
Description
IgT Monoclonal specifically detects IgT in Fish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
IgT | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
0.1 mL | |
Fish | |
IgG |
Western Blot, ELISA, Immunohistochemistry | |
Z55F8*C3 | |
Western Blot, ELISA, Immunohistochemistry | |
IgZ, Immunoglobulin IgT | |
Ovalbumin-conjugated synthetic peptide | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction