Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ihfB Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144436
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144436 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144436 Supplier Invitrogen™ Supplier No. PA5144436
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ihfB Polyclonal Antibody for Western Blot, ELISA

This protein is one of the two subunits of integration host factor, a specific DNA-binding protein that functions in genetic recombination as well as in transcriptional and translational control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ihfB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene ihfB
Gene Accession No. P0A0U2
Gene Alias himD; hip
Gene Symbols ihfB
Host Species Rabbit
Immunogen Recombinant Neisseria meningitidis serogroup B Integration host factor subunit beta(ihfB) (1-104aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66752941
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.