Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IHH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578392
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578392 Supplier Invitrogen™ Supplier No. PA578392
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted Reactivity: Human (100%), Rat (93%), Pig (97%), Bovine (96%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Ihh encodes a member of the hedgehog family of secreted signaling molecules. Hedgehog proteins are essential regulators of a variety of developmental processes including growth, patterning and morphogenesis. The encoded protein specifically plays a role in bone growth an differentiation. Mutations in this gene are the cause of brachydactyly type A1 which is characterized by shortening or malformation of the phalanges. Mutations in this gene are also the cause of acrocapitofemoral dysplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IHH
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Ihh
Gene Accession No. P97812, Q14623
Gene Alias BDA1; HHG2; HHG-2; Ihh; Indian hedgehog; Indian hedgehog homolog; indian hedgehog protein; Indian hedgehog protein C-product; Indian hedgehog protein N-product; indian hedgehog protein; LOW QUALITY PROTEIN: indian hedgehog protein; Indian hedgehog signaling molecule; Indian hedgehog-like protein
Gene Symbols Ihh
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human IHH. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16147, 3549
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.